Autokit Total Ketone Bodies R1 Set, FUJIFILM Irvine Scientific
Supplier: Wako Life Sciences
415-73301
11001-654CS 622 USD
11001-654
Autokit Total Ketone Bodies R1 Set, FUJIFILM Irvine Scientific
Assays Protein Assays
The Autokit Total Ketone Bodies R1 Set is an in vitro assay for the quantitative determination of total ketone bodies in serum or plasma
  • For research use only
  • Quantitative determination of total ketone bodies
  • In vitro assay
The Autokit Total Ketone Bodies R1 Set is an in vitro assay for the quantitative determination of total ketone bodies [acetoacetone (AcAc) + 3 hydroxybutyrate (3-HB)] in serum or plasma. For Research Use Only. Contents: Thio-NAD 2 x for 27 mL, Buffer 2 x 27 mL.