Magid® FiberLock® Nylon Gloves, Magid
Supplier: Magid Glove
31NYXL
11000-625PK 59.59 USD
11000-625 11000-620 11000-622
Magid® FiberLock® Nylon Gloves, Magid
Gloves
These Magid's FiberLock® gloves are ideal for use in restrictive areas in electronics, semiconductor, and automotive paint industries.
  • Manufactured in cleanroom setting
  • Pre-laundered to retain size
  • Good in areas where lint is a problem
Highly reusable, it features a reversible pattern and knit wrists. The gloves are pre-laundered to ensure that they retain their size after washing and the the glove has a high density knit for less snag and penetration.
Certifications: Manufactured in an ISO level 6-rated cleanroom, Class 1000.
Packaging: Packed in a BHT-free polybag.