Magid® TouchMaster® 650H/651H Lightweight Cotton Lisle Inspection Glove, Magid
Supplier: Magid Glove
650H 651H
11000-252CS 1510.42 USD
11000-252 11000-296
Magid® TouchMaster® 650H/651H Lightweight Cotton Lisle Inspection Glove, Magid
Gloves
The inspection glove is 100% cotton two-piece lisle glove constructed of cool and breathable cotton.
  • Two-piece cotton construction
  • Cut and sewn pattern
  • Color-coded sizes for easy sorting
  • Hemmed edge to prevent unraveling
This glove is form fitting and it features a hemmed edge to prevent unraveling in the wash. The glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture. This glove is a great general-purpose glove for light jobs in any industry.