Magid® Kevlar® High Heat Glove, Magid
Supplier: Magid Glove
GG1314WLLP
11000-285CS 1396.15 USD
11000-285
Magid® Kevlar® High Heat Glove, Magid
Gloves
The Magid is extra-heavyweight woven Kevlar glove made to protect your hands while working in extreme temperatures.
  • Extra-heavyweight fabric
  • Wool lining for added thermal protection, comfort
  • 14" long for protection past the wrist
  • One size fits all
This durable high heat resistant gloves feature a high tensile strength that performs well in applications up to 600 to 800 °F. This heavy-duty style comes with a sewn-in wool liner for added thermal protection and comfort.