Wool-Lined Gloves and Mitts, Kevlar® Fiber, Magid
Supplier: Magid Glove
GG1314WL
11000-276EA 67.09 USD
11000-276 11000-266
Wool-Lined Gloves and Mitts, Kevlar® Fiber, Magid
Gloves
Magid® extra-heavyweight woven Kevlar glove made to protect your hands while working in extreme temperatures. Kevlar® 100% wool lined mitts are abrasion, cut and heat resistant. Extra wide cuff opening for easy on and off.
  • Performs well in applications up to 800 °F
  • Wool lining for added thermal protection, comfort
  • 14" long for protection past the wrist
These durable high heat resistant gloves feature a high tensile strength that performs well in applications up to 600 to 800 °F. In addition to its heat-resistant properties, the gloves for heat give you the cut, abrasion protection, and long wear that you've come to know and expect from Kevlar. This heavy-duty style comes with a sewn-in wool liner for added thermal protection and comfort.