Magid® TerryMaster Loops-Out Heavyweight Terrycloth Glove, Magid
Supplier: Magid Glove
PT930R
11000-246EA 10.88 USD
11000-246
Magid® TerryMaster Loops-Out Heavyweight Terrycloth Glove, Magid
Gloves
Magid® TerryMaster loops-out heavyweight terrycloth gloves are seamless, machine knit gloves designed for hard core protection.
  • Economical alternative to hot mill gloves
  • Extra-heavyweight terrycloth glove
  • Excellent heat and thermal protection
This glove features a 30 oz. extra heavyweight blend of cotton and polyester machine-knitted into sturdy, loops out terrycloth. Terrycloth provides the exceptional cut, abrasion, and thermal protection.
Professional testing rates this glove at ANSI cut level 2, giving the PT930R a similar level of protection to aramid and hppe fibers, but for a fraction of the cost.