Eosin Y alcoholic solution molecular biology stain

Supplier: VWR International
Danger

10863-274
10863-274CS 112.02 USD
10863-274
Eosin Y alcoholic solution molecular biology stain
Eosin Y

This fast-acting, alcoholic counterstain stains erythrocytes, collagen, and the cytoplasm of muscle (epithelial cells) different shades of pink


  • Stains in pink
  • Provides an excellent delineation of cytoplasmic components


At least three different shades of pink show different intensities of erythrocytes, collagen, and the cytoplasm of muscle or epithelial cells.

Formula: C₂₀H₈Br₄O₅
MDL Number: MFCD00036189
CAS Number: 15086-94-9
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR