Air Canister Tripod, Restek
Supplier: Restek
24151
10863-240EA 556.12 USD
10863-240
Air Canister Tripod, Restek
Tripods
Restek’s air canister tripod holds two canisters simultaneously for collocated ambient air sampling.
  • Lightweight (12 lbs.) and compact for easy storage and transport
  • Extends from 6 to 9' high
  • Large base provides enhanced stability without additional supports
  • Sturdy, rugged metal design for outdoor sampling and transport
The custom-designed bracket holds most 1, 3, and 6 L canisters securely without any tools.
Ordering information: Air sampling canisters sold separately.