Aura™ Personal Air Sampler Kits, Restek
Supplier: Restek
26485 26484
10862-830KT 1678.1 USD
10862-830 10863-216
Aura™ Personal Air Sampler Kits, Restek
Samplers Air Samplers
New personal air sampling technology designed to meet OSHA and NIOSH requirements for industrial hygiene applications.
  • Quiet, robust sampler
  • Easy to operate
  • Broader utility than analyte-specific sorbent techniques
  • Wider effective range than sorbent tubes and badges
  • Green technology
  • Fully adjustable for a comfortable, secure fit
  • Downloadable XLS file simplifies flow calculation.
Quiet, robust sampler does not require a pump and manages variations in face velocity, temperature, and humidity better than traditional sampling approaches. A simple, quick connection starts and stops flow; does not require flow calibration. Whole-air sampling approach affords multiple analyses of over 100 VOCs. Broader utility than analyte-specific sorbent techniques. Wider effective range than sorbent tubes and badges. Sensitive down to pptv levels and not subject to sample breakthrough even at ppmv levels. Green technology does not use electricity or solvents.