PUF Glass Holders, Restek
Supplier: Restek
22965
10862-926EA 58.65 USD
10862-926 10863-058
PUF Glass Holders, Restek
Samplers Air Samplers
Durable and reusable, PUF glass holders reduce waste and a cost-effective alternative to precleaned packed cartridges.
  • Fit either 6 cm or 22 mm O-Ø PUF plug or can be used with bulk SDVB resin
  • Large glass holder fitted with double stainless steel screens for support
  • Small glass holder has stem designed for secure 1/4" I-Ø tubing connection to sampling pump