TO-Can® Air Sampling Canisters with Swagelok® SS4H Bellows Valve, Restek
Supplier: Restek
22108 22105 22107 22106
10862-934EA 1858.97 USD
10862-934 10862-942 10862-944 10863-230
TO-Can® Air Sampling Canisters with Swagelok® SS4H Bellows Valve, Restek
Samplers Air Samplers
Restek offers a complete line of TO-Can® canisters (SUMMA can equivalent), electropolished using a proprietary process and extensively cleaned using an ultrasonic method.
  • Proprietary electropolished surface that maintains compound stability
  • High quality, Bellows-design valve
  • Now also available with new RAVE™ valve
U.S. EPA Methods TO-14A and TO-15 regulate the collection, storage, and analysis of volatile organic compounds (VOCs) using treated air sampling canisters. This ensures a high-quality, passivated surface that maintains the stability of TO-14A/TO-15 compounds during storage. The frame surrounds the electropolished canister, eliminating the need for weld marks on the sphere, thereby preventing active sites on the canister. The unique holder attaches the handle and base to the canister without welds and protects the canister, tube stub, and valve. Each canister is slightly pressurized with contaminant-free nitrogen prior to shipment.
Ordering information: Supplied with 0–15 psig outlet pressure gauge, brass CGA 170 nut and nipple, and 1/8-inch compression outlet. Do not exceed canister maximum pressure of 40 psig (2.75 bar).