MTS-32 TD Multiple Tube Sequential Sampler, Restek
Supplier: Restek
26464
10862-932EA 17648 USD
10862-932
MTS-32 TD Multiple Tube Sequential Sampler, Restek
Samplers Air Samplers
The MTS-32 TD Multiple Tube Sequential Sampler allows sequential sampling of up to 32 thermal desorption tubes (3.5" each).
  • Worry-free, independent operation lets you get more done
  • Simply set up the unit in the field for extended sampling
  • Retrieve it at the end of the sampling period
  • Reliably collect accurate samples
  • Diffusion-locking technology prevents sample losses or the ingress of contaminants
  • Easy to use and portable
  • Use almost anywhere
  • Sample pollutants in ambient air or continuously monitor industrial emissions