Humidification Chamber, Restek
Supplier: Restek
24282
10862-928EA 372.07 USD
10862-928
Humidification Chamber, Restek
Samplers Air Samplers
Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi.
  • Withstands high pressures
  • Acrylic
  • Easy connection to pressure lines
When cleaning SilcoCan® or TO-Can® canisters, it is important to use humidified air or nitrogen to help remove volatile organic contaminants. Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi. The 1/4-inch inlet and outlet compression fittings allow easy connection to pressure lines on your cleaning system. Our humidification chamber also has an easy-to-open lid for filling with water.