Spectra Gas 7621 High-Purity VOC Regulators, Restek
Supplier: Restek
21572
10862-704EA 2070.39 USD
10862-704
Spectra Gas 7621 High-Purity VOC Regulators, Restek
Gas Regulators
Spectra Gas 7621 High-Purity VOC Regulators with two pressure gauges.
  • Single-stage, stainless steel
  • Two pressure gauges and CGA-180 fitting
  • Stainless steel diaphragm and Kel-F® seat
  • Accurate pressure control even at low flow rates