VWR® Boston Round Bottles, Narrow Mouth, Clear and Amber
Supplier: VWR International
10861-752 10861-754 10861-758 10861-760 10861-762 10861-764 10861-768 10861-770 10861-774 10861-776 10861-778 10861-788 10861-790 10861-796 10861-798 10861-800 10861-802 10861-804 10861-806 10861-808 10861-810 10861-812 10861-814 10861-816 10861-818 10861-820 10861-848 10861-850 10861-852 10861-856 10861-858 10861-860 10861-862 10861-866 10861-868 10861-870 10861-872 10861-874 10861-876 10861-882 VWR89042-710 10861-886 10861-888 10861-894 10861-896 10861-898 10861-900 10861-914 10861-916 10861-922 10861-944 10861-946 10861-948 10861-998 10862-000 10862-490 10862-500 10862-502 10862-504 10862-508 10862-510 10862-514 76329-568 76378-340 10861-767 76419-674 10861-793 10861-881
10861-752CS 158.62 USD
10861-752 10861-754 10861-756 10861-758 10861-760 10861-762 10861-764 10861-768 10861-770 10861-772 10861-774 10861-776 10861-778 10861-788 10861-790 10861-796 10861-798 10861-800 10861-802 10861-804 10861-806 10861-808 10861-810 10861-812 10861-814 10861-816 10861-818 10861-820 10861-848 10861-850 10861-852 10861-856 10861-858 10861-860 10861-862 10861-866 10861-868 10861-870 10861-872 10861-874 10861-876 10861-882 10861-884 10861-886 10861-888 10861-894 10861-896 10861-898 10861-900 10861-914 10861-916 10861-922 10861-944 10861-946 10861-948 10861-998 10862-000 10862-490 10862-492 10862-498 10862-500 10862-502 10862-504 10862-508 10862-510 10862-514 76329-568 76378-340 10861-767 76419-674 10861-793 10861-881
VWR® Boston Round Bottles, Narrow Mouth, Clear and Amber
Bottles Glass Bottles
VWR® Brand Boston Round Bottles are made from soda-lime glass that conforms to USP Type III requirements.
  • Clear or amber coloring
  • Available in a variety of pack sizes
  • Narrow mouth
Bottles are available in clear or amber formats.
Several packaging options are offered for this line of Boston Round Bottles. Bottles are available in a convience pack with closures attached to help maintain cleanliness, they are available in a bulk pack with unattached caps in bags, they are available in a bulk pack without caps, or they are available in a bulk pack with caps attached.