Staticide® LF50 Heavy-Duty Lint-Free Wipes, ACL
Supplier: ACL Staticide
LF50
10861-302CS 730.86 USD
10861-302
Staticide® LF50 Heavy-Duty Lint-Free Wipes, ACL
Wipes Reusable Cleaning Cloths Cloths for General Applications
Staticide® LF50 wipes are made of 45% polyester and 55% wood pulp.
  • Quick and easy cleaning of workshop spills
  • Ideal replacement for paper towels and rags
Heavy-Duty Lint-Free Wipes are made from a polyester/cellulose material that contains no contaminating dyes or binders. Highly absorbent with soft strength, this low particle count wipe holds liquid without breaking down or causing lint contamination.
Ideal for a wide range of cleansing purposes, from everyday general cleaning to use in ESD-safe environments for applying cleaners and chemicals.