Eppendorf® Easypet® 3 Electronic Pipette Controller
Supplier: Eppendorf
4430000018
10860-782EA 676.37 USD
10860-782
Eppendorf® Easypet® 3 Electronic Pipette Controller
Pipettes Pipette Controllers
Electronic pipetting doesn't come any more advanced than the Easypet 3 from Eppendorf. Designed to work with Eppendorf serological pipettes, you get complete speed control and utmost precision in an extremely light and well-balanced body that fits comfortably in your hand, for fatigue-free pipetting.
  • Intuitive and convenient speed adjustment simply done with the tips of your fingers
  • Lightweight, well-balanced and ergonomic design that allows for fatigue-free pipetting
  • Vibrant backlit LEDs provide optical feedback of the remaining battery life
  • Lithium polymer rechargeable battery offers long cordless runtime
  • Smooth setting of pump speed
  • Operation while recharging is possible
  • Autoclavable pipette adapter for sterile applications
  • Quick release of aspirating cone for easy exchange of membrane filters
The lightweight, well-balanced Easypet 3 has been designed considering all ergonomic findings to fit comfortably in your hand and matches its shape to allow for fatigue-free pipetting.
Electronic pipette controller with air cushion system with autoclavable pipette adapter and aspirating cone.
Ordering information: The Easypet® 3 Pipette Controller is supplied with mains/power supply, wall mounting device, shelf stand, and two 0.45 µm membrane filters.