Equivalence C18, HPLC Columns
Supplier: HICHROM LIMITED
EQV-5C18-1546 EQV-5C18-2546 EQV-3C18-GD EQV-5C18-GD EQV-3C18-0546 EQV-3C18-1046 EQV-3C18-1546 EQV-3C18-1002 EQV-3C18-0346 EQV-3C18-7546 EQV-5C18-1504 EQV-5C18-2504 EQV-3C18-1203 EQV-3C18-1503 EQV-5C18-1003 EQV-5C18-1203 EQV-3C18-1502 EQV-5C18-1002 EQV-5C18-0546 EQV-5C18-7546 EQV-5C18-1046 EQV-5C18-1246 EQV-3C18-GD2
10829-196EA 590.19 USD
10829-196 10829-198 10829-200 10829-202 10829-204 10829-206 10829-208 10829-210 10829-212 10860-676 10860-680 10860-682 10860-684 10860-686 10860-688 10860-690 10860-692 10860-694 10860-696 10860-698 10860-700 10860-702 10860-704 10860-706 10860-708 10860-710 10860-712 10860-714 10860-716 10860-718 10860-720 10860-722 10860-724 10860-726 10860-728 10860-730 10860-732 10860-734 10860-736 10860-738 10860-740 10860-742 10860-744 10860-746 10860-748 10860-750 10860-752 10860-754 10860-756 10860-758 10860-760 10860-762 10860-764 10860-766 10860-768 10860-770 10860-772 10860-774 10860-912 10860-914 10860-916 10860-918 10860-920 10860-922 10860-924 10860-926 10860-928 10860-777 10860-779
Equivalence C18, HPLC Columns
Chromatography Columns
Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
  • Excellent efficiency and reproducibility
  • Available in 3 or 5 µm particle sizes
Maximum recommended pH: 2 to 8.