PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
Supplier: New Pig
DRM1059
10860-600EA 310.93 USD
10860-600
PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
Drum Storage
Straight sidewall, UN rated open top drum has ribs for extra stength and allows broader use of drum grabs and other equipment.
  • Straight sidewalls allow broader use of drum grabs, parrot beak lifters and other equipment that can't move tapered drums
  • Ribs add strength and rigidity, UN Rated for storage and transport of solid hazardous materials
  • High-density polyethylene won't rust or corrode
  • Quick-release lever lock band can be wire sealed to deter unauthorized access
Compatible with a wide range of liquids and chemicals.