N-Succinimidyl Acrylate ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

S1722-5GM S1722-25GM
10860-560EA 177.05 USD
10860-560 10860-562
N-Succinimidyl Acrylate ≥98%
N-Succinimidyl Acrylate

Storage temperature - FREEZE: Keep tightly closed.


GHS hazard : Exclamation mark


Caution: Storage temperature - FREEZE: Keep tightly closed.

Formula: C₇H₇NO₄
Storage Temperature: Freezer
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Order Now

Specification Test Results

Purity (GC) 98 %
Melting Point 70.0°-72.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR