N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Storage temperature - LIGHT SENSITIVE, AIR SENSITIVE, MOISTURE SENSITIVE, HEAT SENSITIVE: Keep tightly closed in light-resistant containers.
GHS hazard : Exclamation mark;Health hazard
Caution: Storage temperature - LIGHT SENSITIVE, AIR SENSITIVE, MOISTURE SENSITIVE, HEAT SENSITIVE: Keep tightly closed in light-resistant containers.
Formula:
C₈H₁₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00059008 CAS Number: 3817-11-6 |
Specification Test Results
Purity (GC) | 90.0 % min. |
Specific Gravity (20/20) | 1.0060 - 1.0140 |
Refractive Index (N20/D) | 1.4700 - 1.4740 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...