Leak Diverter for Spraying Pipes, PIG®
Supplier: New Pig
TLS147-YW
10860-096EA 218.02 USD
10860-096
Leak Diverter for Spraying Pipes, PIG®
Spill Control Materials
Material wraps completely around pipe to capture and contain sprays, drips and accumulated condensation.
  • Non-permeable tarp
  • Resists punctures, UV and mildew for long life
  • Quick and easy installation
Non-permeable tarp is vinyl-coated for strength and durability; resists punctures, UV and mildew for long life. Tapered design helps funnel liquid toward center for better drainage. Hook-and-loop fasteners make installation quick and easy and secures diverter in place.