BT Lab Systems Rocker, 3D, G-Biosciences
Supplier: G-Biosciences
BT906
10856-502EA 817.68 USD
10856-502
BT Lab Systems Rocker, 3D, G-Biosciences
Shakers and Mixers Rotating/Rocking Shakers
Rocker, 3D, an economical gyratory rocker with perfect 3-dimensional motion for gentle but thorough mixing.
  • Tube Capacity: 60 x 1.5ml, 30 x 3ml, 21 x 7ml, 17 x 15ml
A 3D gyratory rocker that is an ideal choice for small sample mixing
The concave pad can firmly keeps the tubes on the rocker and is safe for use in cold rooms and incubators.
Suitable for a large selection of tubes, including blood collection tubes and centrifuge tubes. Ideal rocker for all electrophoresis and blotting related applications such as gel staining and destaining.