BT Lab Systems Rocking Shaker, G-Biosciences
Supplier: G-Biosciences
BT907
10856-392EA 1316.84 USD
10856-392
BT Lab Systems Rocking Shaker, G-Biosciences
Shakers and Mixers Rotating/Rocking Shakers
Rocking Shaker is compactly designed, takes up very little space. It is also used for cell culture and cell membrane transfer.
  • Platform made of stainless steel and natural rubber, which is alkali­ resistant, non deforming, easy-clean and spill-prevention
  • Speed real-time display and diminishing time display
  • The bearing is articulated rotating, secure and no wear, suitable for prolonged use
  • Brushless DC motor, maintenance-free
Rocking Shaker is used in the immobilization of running gels, shaking for dyeing or fading Coomassie blue, immobilization, dyeing and developing of silver-staining, developing and fixation of X-ray film in autoradiography experiments, and further processing of cellulose membrane after gel shift such as molecular hybridization or staining with antigen-antibody.
A benchtop shaker suitable for ambient and incubator incubations.