Inserts for 2.0 ml, 8 mm Screw-Thread Vials, Restek
Supplier: Restek
21775 24512 24508 24510 24511 24691
RK21775PK 903.91 USD
RK21775 10803-360 10853-972 10853-974 10853-978 10854-286
Inserts for 2.0 ml, 8 mm Screw-Thread Vials, Restek
Vials
A variety of inserts compatible with screw-thread vials are available in a number of formats, including a flat bottom or with or without a bottom spring.
  • Screw-thread vial compatibility
  • 8 mm orifice
  • Glass or polypropylene fabrication
Inserts for 8 mm screw-thread vials are constructed of borosilicate glass or heavy-duty polypropylene for durable use.