Outlet Cap and Gold Seal Assembly Tool for Agilent 1050, 1100 HPLC Systems, Restek
Supplier: Restek
24989
10853-792EA 122.6 USD
10853-792
Outlet Cap and Gold Seal Assembly Tool for Agilent 1050, 1100 HPLC Systems, Restek
Closures Safety Caps and Safety Waste Caps
Easily install the gold seal into the outlet cap.
Ordering information: For use with Agilent 5001-3701 gold disk seal only. Not compatible with Restek cat.# 25889 and 25890 (Agilent 5067-4728).