Seal Pack Rebuild Kit (without Seal Wash Tube Assembly) for Waters HPLC Systems, Restek
Supplier: Restek
25495
10853-646EA 481.7 USD
10853-646
Seal Pack Rebuild Kit (without Seal Wash Tube Assembly) for Waters HPLC Systems, Restek
Chromatography Supplies Chromatography Spare Parts
Delivery information: Kit includes wash tube seal, seal wash tube, PTFE washer, filter retainer, lower wash seal frit, needle wash frit, TFE washer, needle, injector seal (2), compression screw, ferrule.