Ternary Tee Mixer, Restek
Supplier: Restek
25138
10853-382EA 1053.73 USD
10853-382
Ternary Tee Mixer, Restek
Shakers and Mixers Vortex Mixers
An efficient cross-flow shearing mechanism and interchangeable cartridges produce vortex shear mixing over a wide range of flows.
  • Reduced baseline noise
  • Increased sensitivity
  • Improved gradient accuracy for more reproducible results
  • Increased reaction efficiency in post-column derivatization
Stainless steel.