Tissue-Tek® FormaGO® Formalin Neutralizer Container, Sakura® Finetek
Supplier: Sakura Finetek
9159
10852-360EA 225.31 USD
10852-360
Tissue-Tek® FormaGO® Formalin Neutralizer Container, Sakura® Finetek
Spill Control Materials
The Tissue-Tek® FormaGO® Formalin Neutralizer Container features a narrow opening to reduce exposure to fumes
This empty 9.5L (2.5gal.) container has an integrated handle to facilitate transport.