SHOWA S-TEX300 A4 Cut Resistant, Natural Rubber Coated, Kevlar/Hagane Steel Glove, Showa
Supplier: Showa
S-TEX300S-07 S-TEX300M-08 S-TEX300L-09 S-TEX300XL-10
10852-318CS 2018.41 USD
10852-318 10852-320 10852-322 10852-324
SHOWA S-TEX300 A4 Cut Resistant, Natural Rubber Coated, Kevlar/Hagane Steel Glove, Showa
Gloves
Unique Hagane steel component makes glove extremely cut resistant while maintaining flexibility.
  • Hagane steel reinforced Kevlar construction
  • Tough natural rubber coating
These gloves are used in a wide range of cut resistant applications.
Certifications: Meets ANSI Level A4 cut level.