MXT®-20 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70320
10851-668EA 577.95 USD
10851-668
MXT®-20 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-20 polymer is synthesized to exacting standards so that all residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight mono-modal distribution and extremely low bleed.
  • General-purpose columns for volatile compounds, flavor compounds, and alcoholic beverages
  • Temperature range: −20 to 340 °C
  • Equivalent to USP G28, G32 phases
  • Low to mid-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
MXT®-20 columns are made of Siltek®-treated stainless steel and feature extremely low bleed levels.