MXT®-624 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70971 70968 70969 70973
RK70971EA 1059.67 USD
RK70971 10851-616 10851-618 10851-620
MXT®-624 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
The unique polarity of MXT®-624 columns makes them ideal for analyses of volatile organic pollutants.
  • Application-specific columns for volatile organic pollutants
  • Recommended in U.S. EPA methods for volatile organic pollutants
  • Temperature range: −20 to 280 °C
  • Equivalent to USP G43 phase
  • 4.5" standard coil diameter
  • Low- to mid-polarity phase
Although the MXT®-502.2 column is recommended in many methods, MXT®-624 columns offer the best separation of the early-eluting gases.