MXT®-Volatiles Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
70924 70925 70926 70927
10851-488EA 993.79 USD
10851-488 10851-490 10851-492 10851-494
MXT®-Volatiles Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-Volatiles columns were the first columns designed specifically for analyses of the 34 volatile organic pollutants listed in U.S.
  • Application-specific columns for volatile organic pollutants
  • Temperature range: −20 to 320 °C
  • 4.5" standard coil diameter
  • Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase
With these columns, you can quantify all compounds listed in these methods, whether you use a mass spectrometer or a PID in tandem with an ELCD. The diphenyl/dimethyl polysiloxane based MXT®-Volatiles stationary phase provides low bleed and thermal stability to 320 °C.
EPA methods 601, 602, and 624.