Rtx®-1614 Columns (Fused Silica), Restek
Supplier: Restek
10295 10296
10850-176EA 867.44 USD
10850-176 10850-178
Rtx®-1614 Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-1614 columns feature a unique deactivation that imbues the columns with greater analytical sensitivity.
  • Short column option resolves BDE-209 3 times faster, with less thermal breakdown
  • Unique deactivation gives higher BDE-209 response than competitor columns, for greater analytical sensitivity
  • Exceeds EPA method 1614 resolution criteria for BDE-49 and BDE-71
  • Stable to 360 °C
  • 5% diphenyl, 95% dimethyl polysiloxane
These fused silica columns are optimized for PBDE analysis by EPA method 1614.