Tenax® Packing Materials, Restek
Supplier: RESTEK CORP
Using thermal desorption techniques such as purge-and-trap and short path thermal desoprtion systems, this porous resin can effectively pack columns for the purpose of trapping volatiles and semi-volatiles.
- Stable upto 350 °C
Tenax® can be applied both as a column packing material and for traps for organic volatile and semi-volatile compounds.
Ordering information: Minimum order of 10 grams. Price is per gram.