HayeSep® Packing Materials, Restek
Supplier: Avantor
25032 25033 25036 25037 25038 25039 25045 25046 25047 25048 25050 25054 25055 25056 25563 25568 25569 25570 25571 25572 25574 25575
10847-852EA 240.15 USD
10847-852 10847-858 10847-860 10847-866 10847-868 10847-870 10847-872 10847-878 10847-880 10847-882 10847-884 10847-886 10847-888 10847-890 10847-894 10847-896 10847-898 10847-900 10848-014 10848-024 10848-026 10848-028 10848-030 10848-032 10848-034 10848-036 10848-038
HayeSep® Packing Materials, Restek
Chromatography Sorbents GC Sorbents
HayeSep® porous polymers are macroporous, spherical, ultrapure resins for performing unique separations in gas chromatography.
  • Thoroughly cleaned and preconditioned
  • No shrinkage
  • Low bleed
Columns packed with HayeSep® polymers produce lower bleed and more reproducible characteristics than those packed with any competing polymeric product, and they require minimal conditioning.