Kimtech™ A5 Sterile Hood Over Respirator, Kimberly-Clark Professional
Supplier: Kimberly-Clark
40616
10847-814CS 723.69 USD
10847-814
Kimtech™ A5 Sterile Hood Over Respirator, Kimberly-Clark Professional
Disposable Garments
Kimtech™ A5 sterile elasticized hoods provide an uninterrupted seal and customized fit for those who wear a full-face respirator.
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms
  • Individually bagged hoods have a sterility indicator and are folded inside out for aseptic donning
  • Stretch-fit hood and opening provides an uninterrupted seal around the respirator; great for small faces
  • Clean-Don™ technology ties for easy donning and a more secure fit
  • Manufactured in an ISO-certified facility, clean-processed and individually packaged under cleanroom conditions
Kimtech™ A5 sterile elasticized hoods feature a stretch fit on the hood and opening, with tunneled overseams to prevent particle shedding. These hoods have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. They are made without latex and are designed for single-use (disposable PPE).
Certifications: CE certified.