WHEATON® French Square Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W217883
10847-498CS 228.2 USD
10847-498 10847-502 10847-504 10847-506
WHEATON® French Square Bottles, DWK Life Sciences
Bottles Glass Bottles
High-quality bottles are made from clear USP Type III soda-lime glass.
  • Ideal for solvent, chemical, or sample storage
  • Square shape maximizes storage space
  • Available with caps attached or bulk packs without caps
These are ideal for field use or general laboratory use.