FlexTech™ Seamless Knit Palm Coated Glove, Sandy Nitrile Palm, Wells Lamont®
Supplier: Wells Lamont
Y9216XS Y9216S Y9216M Y9216L Y9216XL Y9216XXL
10847-276PK 300.62 USD
10847-276 10847-278 10847-296 10847-298 10847-300 10847-302
FlexTech™ Seamless Knit Palm Coated Glove, Sandy Nitrile Palm, Wells Lamont®
Gloves
The seamless, machine-knit shell of these gloves provides all-day comfort and protection.
  • Sandy nitrile palm coating for grip in wet and oily conditions
  • Washing machine friendly
  • Cut resistant
  • Applications in metal handling, metal stamping, and automotive manufacturing
Certifications: ASTM F1790-97. ANSI Cut 5.
Ordering information: Gloves are sold by the dozen pairs.