KLEENGUARD® A20 Breathable Particle Protection Bouffant Cap, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
66829
10843-053CS 262.69 USD
10843-053
KLEENGUARD® A20 Breathable Particle Protection Bouffant Cap, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
Disposable bouffant caps are designed to provide protection from submicron sized particles (0.26µm), dry particulates, and light liquid sprays
Lightweight and breathable, caps are best suited for use in cleanroom applications. 24" - Large. Latex-free. Supplied double bagged. NPFA 99 Compliant Antistatic Material.