KIMBLE® KIMAX® Ramsbottom Coking Bulb, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
896650-0025
10842-804CS 395.28 USD
10842-804
KIMBLE® KIMAX® Ramsbottom Coking Bulb, DWK Lifesciences
Petroleum Testing Apparatus
The Ramsbottom Coking Bulb is used in the determination of carbon residue in petroleum products.
Certifications: Ref: ASTM D524. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.