Quercetin dihydrate ≥95%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Q2150-100GM
Q2150-2.5KG
Q2150-25GM
Q2150-500GM
10841-964EA
306.1
USD
10841-964
10841-966
10841-968
10841-970
Quercetin dihydrate ≥95%
Quercetin dihydrate
Quercetin, Dihydrate, also known as meletin, is a flavonol found in several types of vegetables, fruits, leaves and grains.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₅H₁₄O₉ Storage Temperature: Ambient |
MDL Number:
MFCD00149487 CAS Number: 6151-25-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay (C₁₅H₁₀O₇.2H₂O) | 95 % min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...