Brady® BBP®85 and PowerMark® Industrial Grade Print Ribbon
Supplier: Brady Worldwide
13511
10841-222EA 275.09 USD
10841-222
Brady® BBP®85 and PowerMark® Industrial Grade Print Ribbon
Printer Ribbons
This industrial-grade ribbon is uniquely designed with memory cells that communicate information to the BBP®85 and PowerMark® sign and label printers.
BBP®85 and PowerMark® Industrial Grade Print Ribbon is super-tough and smear-proof to ensure printed text lasts for years, both outdoors and in industrial areas.