REPEATER Pump Tube Set, Baxter IV Solutions
Supplier: Baxter
H93811
10837-306CS 522.85 USD
10837-306
REPEATER Pump Tube Set, Baxter IV Solutions
Infusions
REPEATER pump tube set with universal spike-luer lock.
  • Sterile
  • Rx only
It consists of inlet spike and male luer lock outlet for transfer of sterile fluids from one container to another using the REPEATER Pump.
Delivery information: Includes RAPIDFILL Connectors, Dispensing Pins or TWOFER Needles.