Water, certified for molecular biology

Supplier: Rocky Mountain Biologicals

WFI-MGW-20L
10837-214EA 290 USD
10837-214
Water, certified for molecular biology
Water

Cytiva Molecular Grade water is quality tested and certified RNase, DNase, and protease free.


  • Purified utilizing a nine step process resulting in very high quality water


Every lot sent to our customer contains zero traces of toxic agents such as DEPC, and is well suited for use in diverse molecular biology applications.


Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Appearance Clear Liquid (Visual)
TOC 500 ppb (USP 643)
Conductivity 5.0 µs/cm (USP 645)
DNAse None Detected
Protease None Detected
RNAse None Detected
GMP manufactured conform


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR