Water USP, purified, sterile

Supplier: Cytiva

WPW-USP-1X6
10837-202CS 148 USD
10837-202 10837-204 10837-206
Water USP, purified, sterile
Water

Cytiva's Purified Water, USP can be used as an excipient in the production of non-parenteral preparations such as cleaning of certain equipment and non-parenteral product-contact components.


  • Sterile
  • Purified utilizing a nine step process resulting in very high quality water


Available in sterile and non-sterile formats, it complies with the USP monograph for purified water.


Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear Liquid (Visual)
Conductivity (Packaged) 5.0 µs/cm (USP 645)
Sterility No Growth (USP 71)
TOC (Before Packaging) 500 ppb (USP 643)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR