OHM Metrics™ Digital Surface Resistance Test Kit, Transforming Technologies
Supplier: Transforming Technologies
SRM500K
10837-050EA 3148.49 USD
10837-050
OHM Metrics™ Digital Surface Resistance Test Kit, Transforming Technologies
Static Control Monitors
The SRM500K is a digital surface resistance test kit designed to test all conductive, anti-static, and static dissipative surfaces for electrical resistivity/resistance according to EOS/ESD, CECC, ANSI, ASTM test procedures
Standard features include 10V and 100V range voltage selection, automatic zeroing, and power shut off. The unit includes two 2.3kg (5lb.) disk probes, built-in resistivity probes, and carrying case. The device offers a test range of 10³ to 10¹². Readout is provided on an LCD alpha-numeric scale, without LEDs.
It accurately and quickly measures resistance between two points (RTT) and surface to ground resistance (RTG). Suitable for factory audits and test lab evaluations, the OHM Metrics™ SRM500K also measures ambient temperature and humidity.