AimTab™ Ketone Tablets, Germaine® Laboratories
Supplier: Germaine Laboratories
13100
10837-000PK 97.06 USD
10837-000
AimTab™ Ketone Tablets, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
AimTab™ Ketone Tablets are desgined for the semi-quantitative determination of ketones (acetoacetic acid and acetone) in urine, serum, plasma, or whole blood
  • Specifically designed for acetoacetic acid and acetone
  • Tablets will not react with betahydroxybutyric
  • CLIA moderately complex when testing serum, plasma, or whole blood
  • Same test procedure as Acetest®
Tablets detect as little as 5mg/dL of acetoacetic acid in urine.
Certifications: CLIA waived for urine.