VWR® Cleanroom Wipes, 100% Polyester
Supplier: VWR International
P3-LG12B
10835-982CS 1385.52 USD
10835-982
VWR® Cleanroom Wipes, 100% Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These are 100% continuous-filament, double-knit polyester fabric wipes.
  • Free from silicone, amides, and DOP contamination
  • Extremely low particle and fiber levels
  • Laser cut edges
  • Absorbant
Resistant to abrasion, providing maximum cleaning performance. Designed for cleaning surfaces and controlling spills in ISO Class 4 cleanroom environments.