Potassium Hydroxide, 20% KOH, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z78
10834-134EA 28.87 USD
10834-134
Potassium Hydroxide, 20% KOH, Hardy Diagnostics
Microbiology Test Systems Enzymatic Tests
For use in slide preparations of specimens for the purpose of identifying fungal elements
  • Slide preparations of specimens
  • Dissolves epithelial cells
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Dissolves epithelial cells that may mask fungal elements.